Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mss51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106025.1 | NP_001099495.1 | 4 - 5 | 561 | 140 | |
GenBank mRNA | FQ215928.1 | - | 2 - 3 | 208 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |