Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012606.2 | NP_036738.1 | 3 - 4 | 375 | 85 | |
XM_006243917.2 | XP_006243979.2 | 3 - 4 | 476 | 85 | ||
GenBank mRNA | BC081832.1 | - | 3 - 4 | 375 | 7192037 | 85 |
FQ216106.1 | - | 3 - 4 | 387 | 85 | ||
FQ216251.1 | - | 3 - 4 | 387 | 85 | ||
FQ216255.1 | - | 3 - 4 | 407 | 85 | ||
FQ217222.1 | - | 3 - 4 | 390 | 85 | ||
FQ223547.1 | - | 3 - 4 | 387 | 85 | ||
X14812.1 | - | 3 - 4 | 368 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |