Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053999.2 | NP_446451.1 | 5 - 6 | 762 | 109 | |
XM_008770730.1 | XP_008768952.1 | 5 - 6 | 1569 | 109 | ||
XM_008770731.1 | XP_008768953.1 | 5 - 6 | 1569 | 109 | ||
XM_008770732.1 | XP_008768954.1 | 3 - 4 | 351 | 109 | ||
GenBank mRNA | BC128731.1 | - | 5 - 6 | 762 | 7370343 | 109 |
D14419.1 | - | 2 - 3 | 222 | 109 | ||
M83297.1 | - | 6 - 7 | 708 | 109 | ||
M83298.1 | - | 5 - 6 | 743 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Rattus norvegicus Probe spans exons |