Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021771.2 | NP_068539.2 | 11 - 12 | 2610 | 156 | |
XM_006232282.2 | XP_006232344.2 | 11 - 12 | 2775 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel 3' Most Rattus norvegicus 1700 Probe spans exons |