Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022232.1 | NP_071568.1 | 9 - 10 | 1076 | 97 | |
GenBank mRNA | AB017702.3 | - | 9 - 10 | 1076 | 97 | |
BC061764.1 | - | 9 - 10 | 1171 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |