Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022600.1 | NP_072122.1 | 9 - 10 | 2549 | 108 | |
XM_008768744.1 | XP_008766966.1 | 9 - 10 | 2722 | 108 | ||
GenBank mRNA | M96159.1 | - | 9 - 10 | 2549 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |