Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snap47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199389.1 | NP_955421.1 | 2 - 3 | 626 | 132 | |
XM_006246488.2 | XP_006246550.1 | 2 - 3 | 660 | 132 | ||
GenBank mRNA | BC062056.1 | - | 2 - 3 | 626 | 5598911 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |