Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201350.1 | NP_958738.1 | 1 - 2 | 90 | 135 | |
GenBank mRNA | AY298755.1 | - | 1 - 2 | 90 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |