Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckap2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001057508.5 | XP_001057508.3 | 3 - 4 | 233 | 73 | |
XM_006234995.2 | XP_006235057.1 | 3 - 4 | 233 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |