Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mslnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767615.1 | XP_008765837.1 | 12 - 13 | 3579 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |