Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cables2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006235831.2 | XP_006235893.2 | 8 - 9 | 1106 | 130 | |
XM_008762585.1 | XP_008760807.1 | 8 - 9 | 1115 | 130 | ||
XM_008762586.1 | XP_008760808.1 | 8 - 9 | 1106 | 130 | ||
XM_008762588.1 | XP_008760810.1 | 7 - 8 | 1043 | 130 | ||
XM_008762589.1 | XP_008760811.1 | 8 - 9 | 1115 | 130 | ||
XM_008762590.1 | XP_008760812.1 | 8 - 9 | 1115 | 130 | ||
XM_008762591.1 | XP_008760813.1 | 8 - 9 | 1115 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |