Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011961.1 | NP_001011961.1 | 6 - 7 | 1270 | 78 | |
GenBank mRNA | BC090006.1 | - | 6 - 7 | 1270 | 7323399 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |