Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sugp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108397.1 | NP_001101867.1 | 2 - 3 | 538 | 88 | |
XM_008771124.1 | XP_008769346.1 | 1 - 2 | 125 | 88 | ||
XM_008771125.1 | XP_008769347.1 | 2 - 3 | 165 | 88 | ||
XM_008771126.1 | XP_008769348.1 | 2 - 3 | 300 | 88 | ||
XM_008771127.1 | XP_008769349.1 | 1 - 2 | 125 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |