Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcidas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001068366.4 | XP_001068366.2 | 5 - 6 | 652 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |