Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sft2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034011.1 | NP_001029183.1 | 2 - 3 | 164 | 79 | |
GenBank mRNA | BC099092.1 | - | 2 - 3 | 164 | 7457938 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |