Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107942.2 | NP_001101412.2 | 2 - 3 | 203 | 67 | |
GenBank mRNA | BP504151.1 | - | 2 - 3 | 188 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |