Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc41a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108742.1 | NP_001102212.1 | 8 - 9 | 1620 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |