Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106153.1 | NP_001099623.1 | 2 - 3 | 237 | 74 | |
GenBank mRNA | FQ211751.1 | - | 2 - 3 | 247 | 74 | |
FQ212326.1 | - | 2 - 3 | 213 | 74 | ||
FQ216914.1 | - | 2 - 3 | 215 | 74 | ||
FQ216957.1 | - | 2 - 3 | 232 | 74 | ||
FQ217727.1 | - | 2 - 3 | 211 | 74 | ||
FQ223951.1 | - | 2 - 3 | 212 | 74 | ||
FQ224026.1 | - | 2 - 3 | 215 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |