Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750410.3 | XP_003750458.1 | 6 - 7 | 1201 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |