Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nanos3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105945.1 | NP_001099415.1 | 1 - 2 | 492 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons |