Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hlcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248111.2 | XP_006248173.1 | 6 - 7 | 2742 | 83 | |
XM_006248112.2 | XP_006248174.1 | 6 - 7 | 2809 | 83 | ||
XM_006248114.2 | XP_006248176.1 | 5 - 6 | 2603 | 83 | ||
XM_008768611.1 | XP_008766833.1 | 6 - 7 | 1941 | 83 | ||
XM_008768612.1 | XP_008766834.1 | 5 - 6 | 1806 | 83 | ||
XM_221630.8 | XP_221630.6 | 5 - 6 | 1922 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |