Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170324.1 | NP_001163795.1 | 1 - 2 | 85 | 94 | |
GenBank mRNA | BC160863.1 | - | 1 - 2 | 85 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |