Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aasdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250858.2 | XP_006250920.1 | 12 - 13 | 3002 | 127 | |
XM_008770108.1 | XP_008768330.1 | 8 - 9 | 2162 | 127 | ||
XM_008770109.1 | XP_008768331.1 | 9 - 10 | 2261 | 127 | ||
XM_008770110.1 | XP_008768332.1 | 9 - 10 | 2261 | 127 | ||
XM_008770111.1 | XP_008768333.1 | 9 - 10 | 2261 | 127 | ||
XM_008770112.1 | XP_008768334.1 | 4 - 5 | 2392 | 127 | ||
XM_008770113.1 | XP_008768335.1 | 5 - 6 | 1351 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Probe spans exons |