Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pus7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729847.4 | XP_002729893.2 | 3 - 4 | 1178 | 139 | |
XM_006242271.2 | XP_006242333.1 | 3 - 4 | 1485 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |