Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008322.1 | NP_001008323.1 | 5 - 6 | 654 | 65 | |
XM_006241453.2 | XP_006241515.1 | 5 - 6 | 680 | 65 | ||
GenBank mRNA | BC085331.1 | - | 5 - 6 | 654 | 7106911 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons |