Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009542.2 | NP_001009542.1 | 3 - 4 | 354 | 102 | |
XM_006232501.2 | XP_006232563.1 | 3 - 4 | 315 | 102 | ||
GenBank mRNA | BC067245.1 | - | 3 - 4 | 358 | 6888212 | 102 |
FQ213019.1 | - | 3 - 4 | 355 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |