Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otop2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105851.1 | NP_001099321.1 | 3 - 4 | 545 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |