Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099504.1 | NP_001092974.1 | 3 - 4 | 1310 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Rattus norvegicus Probe spans exons |