Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145755.2 | NP_001139227.2 | 4 - 5 | 2159 | 66 | |
XM_006246755.2 | XP_006246817.1 | 5 - 6 | 2447 | 66 | ||
GenBank mRNA | FJ665183.1 | - | 4 - 5 | 2150 | 66 | |
HM060628.1 | - | 4 - 5 | 2159 | 66 | ||
HM060629.1 | - | 4 - 5 | 2159 | 66 | ||
HM060630.1 | - | 4 - 5 | 2143 | 66 | ||
HM060631.1 | - | 4 - 5 | 2159 | 66 | ||
HM060632.1 | - | 4 - 5 | 2159 | 66 | ||
HM060633.1 | - | 4 - 5 | 2143 | 66 | ||
HM060634.1 | - | 4 - 5 | 2159 | 66 | ||
HM060635.1 | - | 4 - 5 | 2143 | 66 | ||
HM060636.1 | - | 4 - 5 | 2143 | 66 | ||
HM060637.1 | - | 4 - 5 | 2159 | 66 | ||
HM060638.1 | - | 4 - 5 | 2143 | 66 | ||
HM060639.1 | - | 4 - 5 | 2143 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis Detect Genomic DNA Rattus norvegicus Probe spans exons |