Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053446.1 | NP_445898.1 | 1 | 1213 | 119 | |
GenBank mRNA | AF321129.1 | - | 1 | 535 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |