Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009683.1 | NP_001009683.1 | 2 - 3 | 372 | 174 | |
GenBank mRNA | BC087678.1 | - | 2 - 3 | 372 | 7318816 | 174 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |