Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014771.2 | NP_001014771.1 | 2 | 1961 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |