Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024803.1 | NP_001019974.1 | 8 - 9 | 1549 | 109 | |
XM_006252325.2 | XP_006252387.1 | 8 - 9 | 1609 | 109 | ||
XM_006252326.2 | XP_006252388.1 | 8 - 9 | 1643 | 109 | ||
XM_006252327.2 | XP_006252389.1 | 8 - 9 | 1592 | 109 | ||
GenBank mRNA | BC083605.1 | - | 8 - 9 | 1609 | 109 | |
BC097406.1 | - | 8 - 9 | 1549 | 7453994 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |