Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005904.1 | NP_001005904.1 | 1 | 1778 | 100 | |
GenBank mRNA | BC083832.1 | - | 1 | 1778 | 7112736 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |