Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008804.1 | NP_001008804.1 | 4 | 949 | 89 | |
XM_006247391.2 | XP_006247453.1 | 4 | 6846 | 89 | ||
XM_008768091.1 | XP_008766313.1 | 4 | 6846 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |