Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fez2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053600.2 | NP_446052.2 | 4 - 5 | 640 | 78 | |
XM_006239638.2 | XP_006239700.1 | 4 - 5 | 691 | 78 | ||
XM_006239639.2 | XP_006239701.1 | 4 - 5 | 691 | 78 | ||
GenBank mRNA | AB076183.1 | - | 4 - 5 | 640 | 78 | |
AB080086.1 | - | 4 - 5 | 619 | 78 | ||
BC100060.1 | - | 4 - 5 | 621 | 7128839 | 78 | |
CK359698.1 | - | 4 - 5 | 607 | 78 | ||
U64689.1 | - | 4 - 5 | 466 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |