Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thnsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025035.1 | NP_001020206.1 | 4 | 2527 | 107 | |
GenBank mRNA | BC091213.1 | - | 4 | 2527 | 7382276 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon |