Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077680.1 | NP_001071148.2 | 6 - 7 | 637 | 57 | |
XM_006235355.2 | XP_006235417.1 | 6 - 7 | 716 | 57 | ||
GenBank mRNA | BC127515.1 | - | 6 - 7 | 637 | 7100079 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |