Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099482.1 | NP_001092952.1 | 4 - 5 | 1539 | 144 |
Gene Symbol: LOC103689970 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765901.1 | XP_008764123.1 | 2 - 3 | 297 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Rattus norvegicus Probe spans exons |