Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lilrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100123.1 | NP_001093593.1 | 1 - 2 | 91 | 113 | |
XM_006228170.2 | XP_006228232.1 | 1 - 2 | 176 | 113 | ||
GenBank mRNA | AY998483.1 | - | 1 - 2 | 70 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |