Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mir34c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031849.1 | 1 | 38 | 70 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | rCG58364-like | LOC100359479 | Sense | Chr. 8 - 55471705 - 55494857 on NCBI Build Rnor_6.0 | ||
Downstream | microRNA 34b | Mir34b | rno-mir-34b | 442.0 | Sense | Chr. 8 - 55492542 - 55492625 on NCBI Build Rnor_6.0 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |