Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Padi6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191766.1 | NP_001178695.1 | 1 - 2 | 86 | 76 | |
XM_008764232.1 | XP_008762454.1 | 1 - 2 | 136 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |