Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpsf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014245.2 | NP_001014267.1 | 5 - 6 | 993 | 93 | |
XM_006231061.2 | XP_006231123.1 | 7 - 8 | 1175 | 93 | ||
XM_006231062.2 | XP_006231124.1 | 6 - 7 | 1163 | 93 | ||
XM_008760238.1 | XP_008758460.1 | 6 - 7 | 1094 | 93 | ||
XM_008760239.1 | XP_008758461.1 | 6 - 7 | 1067 | 93 | ||
XM_008760240.1 | XP_008758462.1 | 5 - 6 | 981 | 93 | ||
GenBank mRNA | BC083864.1 | - | 5 - 6 | 996 | 7127736 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |