Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017464.2 | NP_001017464.1 | 8 - 9 | 897 | 72 | |
XM_006230403.2 | XP_006230465.1 | 8 - 9 | 997 | 72 | ||
XM_008759903.1 | XP_008758125.1 | 8 - 9 | 997 | 72 | ||
XM_008759904.1 | XP_008758126.1 | 6 - 7 | 663 | 72 | ||
XM_008759905.1 | XP_008758127.1 | 8 - 9 | 997 | 72 | ||
XM_008759906.1 | XP_008758128.1 | 8 - 9 | 997 | 72 | ||
GenBank mRNA | BC079317.1 | - | 8 - 9 | 900 | 7135047 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |