Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 851072 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270756.1 | NP_001257685.1 | 1 | 168 | 77 |
Gene Symbol: RSC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001182246.1 | NP_013461.1 | 1 | 2553 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon |