Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001183561.3 | NP_014785.3 | 1 | 847 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon |