Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205279.1 | NP_001192208.1 | 2 - 3 | 215 | 106 | |
XM_005653290.3 | XP_005653347.1 | 485 | 106 | |||
GenBank mRNA | AK392297.1 | - | 2 - 3 | 298 | 106 | |
AY374470.1 | - | 2 - 3 | 215 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |