Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MADCAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037998.2 | NP_001033087.1 | 4 | 853 | 59 | |
XM_021081941.1 | XP_020937600.1 | 865 | 59 | |||
XM_021081945.1 | XP_020937604.1 | 886 | 59 | |||
GenBank mRNA | DQ288285.1 | - | 4 | 854 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |