Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSCN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044547.1 | NP_001038012.1 | 1 - 2 | 248 | 79 | |
GenBank mRNA | DQ324422.1 | - | 1 - 2 | 248 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |