Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044567.1 | NP_001038032.1 | 3 - 4 | 1058 | 77 | |
XM_021064070.1 | XP_020919729.1 | 1546 | 77 | |||
XM_021064071.1 | XP_020919730.1 | 1479 | 77 | |||
XM_021064072.1 | XP_020919731.1 | 1710 | 77 | |||
XM_021064073.1 | XP_020919732.1 | 1543 | 77 | |||
GenBank mRNA | AK239748.1 | - | 4 - 5 | 1531 | 77 | |
AK393877.1 | - | 2 - 3 | 342 | 77 | ||
DQ450901.1 | - | 3 - 4 | 1058 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Inventoried Probe spans exons |